ENAH2C55717E Enamine

2-{[(tert-butoxy)carbonyl]amino}-3-(3,5-dibromophenyl)propanoic acid

  • MDL number MFCD09916559
  • Cas Number : 1259976-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725155
Availability details for
Quantity
  • Availability
  • Add to Cart