ENAH2C557188 Enamine

2-(1,4-dioxan-2-yl)ethane-1-sulfonyl chloride

  • MDL number MFCD31667671
  • Cas Number : 2091573-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725266
Availability details for
Quantity
  • Availability
  • Add to Cart