ENAH2C55719A Enamine

2-chloro-1-{imidazo[1,5-a]pyridin-1-yl}ethan-1-one

  • MDL number MFCD20628101
  • Cas Number : 51132-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725574
Availability details for
Quantity
  • Availability
  • Add to Cart