ENAH2C5571A1 Enamine

2-(naphthalen-2-yl)-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD31666930
  • Cas Number : 1141931-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725673

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725673
Availability details for
Quantity
  • Availability
  • Add to Cart