ENAH2C5571A9 Enamine

{[2-(trifluoromethyl)phenyl]carbamoyl}methyl (2E)-3-(3-methoxyphenyl)prop-2-enoate

  • MDL number MFCD03976662
  • Cas Number : 728927-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725816
Availability details for
Quantity
  • Availability
  • Add to Cart