ENAH2C5571AF Enamine

benzyl N-(4-aminophenyl)carbamate

  • MDL number MFCD06657916
  • Molecular Weight : 242.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 4-aminophenylcarbamate
Inchi Code 1S/C14H14N2O2/c15-12-6-8-13(9-7-12)16-14(17)18-10-11-4-2-1-3-5-11/h1-9H,10,15H2,(H,16,17)
Inchi Key PITLFTKLRFOUEJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1725883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725883
Availability details for
Quantity
  • Availability
  • Add to Cart