ENAH2C5571B1 Enamine

3-isothiocyanato-2-methoxypyridine

  • MDL number MFCD18803414
  • Cas Number : 52023-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1725900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1725900
Availability details for
Quantity
  • Availability
  • Add to Cart