ENAH2C5571BC Enamine

1,2-benzothiazol-7-amine

  • MDL number MFCD01658891
  • Cas Number : 89795-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726077

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726077
Availability details for
Quantity
  • Availability
  • Add to Cart