ENAH2C5571C1 Enamine

ethyl 3-(chloromethyl)-1,2-oxazole-4-carboxylate

  • MDL number MFCD20637794
  • Cas Number : 1379165-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726124
Availability details for
Quantity
  • Availability
  • Add to Cart