ENAH2C5571C2 Enamine

4-bromo-1-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD28368569

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726140
Availability details for
Quantity
  • Availability
  • Add to Cart