ENAH2C5571C3 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-1H-imidazole-4-carboxylic acid

  • MDL number MFCD23767344
  • Cas Number : 1499626-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726150
Availability details for
Quantity
  • Availability
  • Add to Cart