ENAH2C5571C4 Enamine

2-ethylquinoline-3-carboxylic acid hydrochloride

  • MDL number MFCD26383500
  • Cas Number : 888014-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726156

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726156
Availability details for
Quantity
  • Availability
  • Add to Cart