ENAH2C5571C5 Enamine

3-bromo-5-(methylsulfanyl)-1,2,4-thiadiazole

  • MDL number MFCD02946284
  • Cas Number : 36955-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726161
Availability details for
Quantity
  • Availability
  • Add to Cart