ENAH2C5571CD Enamine

(2E)-3-(5-methoxypyridin-3-yl)prop-2-enoic acid

  • MDL number MFCD20655053
  • Cas Number : 118419-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726220

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726220
Availability details for
Quantity
  • Availability
  • Add to Cart