ENAH2C5571CF Enamine

4-(chloromethyl)-5-methyl-2-[3-(trifluoromethyl)phenyl]-1,3-oxazole

  • MDL number MFCD09759022
  • Cas Number : 678164-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726248

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726248
Availability details for
Quantity
  • Availability
  • Add to Cart