ENAH2C5571D3 Enamine

1-tert-butyl-6-chloro-3-methyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD11890473
  • Cas Number : 1181393-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726276
Availability details for
Quantity
  • Availability
  • Add to Cart