ENAH2C5571D7 Enamine

tert-butyl 5-(2,2,2-trifluoroacetyl)-1,2,3,4-tetrahydropyridine-1-carboxylate

  • MDL number MFCD31667008
  • Cas Number : 1378929-70-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726377
Availability details for
Quantity
  • Availability
  • Add to Cart