ENAH2C5571D8 Enamine

(6-fluoro-1-benzofuran-2-yl)boronic acid

  • MDL number MFCD27921787
  • Cas Number : 1800231-48-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726382

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726382
Availability details for
Quantity
  • Availability
  • Add to Cart