ENAH2C5571DB Enamine

7-bromo-2-(trifluoromethyl)-4H-3,1-benzoxazin-4-one

  • MDL number MFCD31667677
  • Cas Number : 2230803-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726417
Availability details for
Quantity
  • Availability
  • Add to Cart