ENAH2C5571DF Enamine

tert-butyl N-[4-(4-aminophenyl)butyl]carbamate

  • MDL number MFCD24466397
  • Cas Number : 742102-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1726452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1726452
Availability details for
Quantity
  • Availability
  • Add to Cart