ENAH2C5571E0 Enamine

(4-chlorophenyl)(2-fluorophenyl)methanone

  • MDL number MFCD00129781
  • Cas Number : 59396-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1732266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1732266
Availability details for
Quantity
  • Availability
  • Add to Cart