ENAH2C5571E4 Enamine

(4-chlorophenyl)(2-methoxyphenyl)methanone

  • MDL number MFCD01311535
  • Cas Number : 78589-10-3
  • Molecular Weight : 246.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1732270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1732270
Availability details for
Quantity
  • Availability
  • Add to Cart