ENAH2C5571E5 Enamine

(4-chlorophenyl)[3-(trifluoromethyl)phenyl]methanone

  • MDL number MFCD12653585
  • Cas Number : 91503-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1732271

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1732271
Availability details for
Quantity
  • Availability
  • Add to Cart