ENAH2C5571E7 Enamine

3,4-dimethoxythiophene-2-carbonitrile

  • MDL number MFCD31667009
  • Cas Number : 2230798-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1732279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1732279
Availability details for
Quantity
  • Availability
  • Add to Cart