ENAH2C5571EB Enamine

2-(1H-pyrazol-4-yl)-1H-imidazole

  • MDL number MFCD31668196
  • Cas Number : 2243507-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1790045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1790045
Availability details for
Quantity
  • Availability
  • Add to Cart