ENAH2C5571F4 Enamine

3-methylpentane-2,3-diol

  • MDL number MFCD18976470
  • Cas Number : 63521-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007700
Availability details for
Quantity
  • Availability
  • Add to Cart