ENAH2C5571F5 Enamine

ethyl 2-(4-carbamoylphenoxy)-2,2-difluoroacetate

  • MDL number MFCD23241639
  • Cas Number : 1503160-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007708
Availability details for
Quantity
  • Availability
  • Add to Cart