ENAH2C5571F8 Enamine

methyl({[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]sulfamoyl})amine

  • MDL number MFCD31667758

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007755
Availability details for
Quantity
  • Availability
  • Add to Cart