ENAH2C5571F9 Enamine

4H,5H,7H-thiopyrano[3,4-b]furan-4-one

  • MDL number MFCD24698988
  • Cas Number : 6881-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007769
Availability details for
Quantity
  • Availability
  • Add to Cart