ENAH2C5571FA Enamine

1-(4-aminophenyl)-3-methyl-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD31667759
  • Cas Number : 2230799-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007793
Availability details for
Quantity
  • Availability
  • Add to Cart