ENAH2C557200 Enamine

4-methyl-N-[2-(pyridin-2-yl)ethyl]-1,3-thiazol-2-amine

  • MDL number MFCD11117243
  • Cas Number : 1036462-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007846
Availability details for
Quantity
  • Availability
  • Add to Cart