ENAH2C557206 Enamine

5-[(tert-butyldimethylsilyl)oxy]pent-1-en-3-one

  • MDL number MFCD31666947
  • Cas Number : 1629341-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2007911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2007911
Availability details for
Quantity
  • Availability
  • Add to Cart