ENAH2C55720B Enamine

3,3-difluoro-2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride

  • MDL number MFCD28012479
  • Cas Number : 1706454-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008040
Availability details for
Quantity
  • Availability
  • Add to Cart