ENAH2C55720C Enamine

methyl (3S)-3-amino-4-(benzyloxy)butanoate hydrochloride

  • MDL number MFCD29086534
  • Cas Number : 1352824-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008049
Availability details for
Quantity
  • Availability
  • Add to Cart