ENAH2C557210 Enamine

tert-butyl 4-(cyanomethyl)-3,3-difluoropiperidine-1-carboxylate

  • MDL number MFCD31667060
  • Cas Number : 1781075-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008080
Availability details for
Quantity
  • Availability
  • Add to Cart