ENAH2C557211 Enamine

3-bromo-1,1-difluoro-1H,1aH,6H,6aH-cyclopropa[a]indene

  • MDL number MFCD31667760
  • Cas Number : 2230808-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008081

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008081
Availability details for
Quantity
  • Availability
  • Add to Cart