ENAH2C557217 Enamine

(2S,3S)-2-methylpyrrolidin-3-ol hydrochloride

  • MDL number MFCD30726588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008120
Availability details for
Quantity
  • Availability
  • Add to Cart