ENAH2C55721B Enamine

3-(3-bromo-4-methoxyphenyl)-3-{[(tert-butoxy)carbonyl]amino}propanoic acid

  • MDL number MFCD31666948
  • Cas Number : 1698166-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008172
Availability details for
Quantity
  • Availability
  • Add to Cart