ENAH2C55721E Enamine

5-(aminomethyl)-1-ethyl-6-methyl-N-(oxan-4-yl)-1H-pyrazolo[3,4-b]pyridin-4-amine

  • MDL number MFCD31667064
  • Cas Number : 932113-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008186
Availability details for
Quantity
  • Availability
  • Add to Cart