ENAH2C557225 Enamine

(1s,3s)-3-[(1H-pyrazol-1-yl)methyl]cyclobutan-1-amine dihydrochloride

  • Cas Number : 2230789-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008217
Availability details for
Quantity
  • Availability
  • Add to Cart