ENAH2C557229 Enamine

3-methyl-3H-imidazo[4,5-c]pyridine

  • MDL number MFCD13175870

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-97
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008239
Availability details for
Quantity
  • Availability
  • Add to Cart