ENAH2C55722B Enamine

1,3-dicyclopentylpropan-2-one

  • MDL number MFCD31690925
  • Cas Number : 858835-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008253
Availability details for
Quantity
  • Availability
  • Add to Cart