ENAH2C557231 Enamine

3-(2,2-difluoroethyl)piperidine hydrochloride

  • MDL number MFCD31667761
  • Cas Number : 2230802-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008280
Availability details for
Quantity
  • Availability
  • Add to Cart