ENAH2C557232 Enamine

octahydro-2H-1,3-benzoxazine

  • MDL number MFCD24498600
  • Cas Number : 90204-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008285
Availability details for
Quantity
  • Availability
  • Add to Cart