ENAH2C557235 Enamine

(2S)-4-methyl-2-(methylamino)pentan-1-ol

  • MDL number MFCD19217154
  • Cas Number : 10203-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008434
Availability details for
Quantity
  • Availability
  • Add to Cart