ENAH2C557238 Enamine

2-chloro-4-(2-methoxyethoxy)pyrimidine

  • MDL number MFCD16712380
  • Cas Number : 110821-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008444
Availability details for
Quantity
  • Availability
  • Add to Cart