ENAH2C557239 Enamine

6-amino-2,3-dihydro-1lambda6,2-benzothiazole-1,1-dione

  • MDL number MFCD18533504
  • Cas Number : 916438-46-5
  • Molecular Weight : 184.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dihydro-1,2-benzisothiazol-6-amine 1,1-dioxide
Inchi Code 1S/C7H8N2O2S/c8-6-2-1-5-4-9-12(10,11)7(5)3-6/h1-3,9H,4,8H2
Inchi Key MJRBDTGIFOARNW-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-2008450

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-2008450
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart