ENAH2C557257 Enamine

1-(4-bromo-2-methoxyphenyl)methanamine hydrochloride

  • MDL number MFCD11045220
  • Cas Number : 2230802-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008645
Availability details for
Quantity
  • Availability
  • Add to Cart