ENAH2C55725A Enamine

sodium 2-[1-(hydroxymethyl)cyclohexyl]acetate

  • MDL number MFCD01734454
  • Cas Number : 7009-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008661
Availability details for
Quantity
  • Availability
  • Add to Cart