ENAH2C55725B Enamine

ethyl 5-(3-chloro-4-methylthiophen-2-yl)-1,3-oxazole-4-carboxylate

  • MDL number MFCD31691339
  • Cas Number : 1969024-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2008668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2008668
Availability details for
Quantity
  • Availability
  • Add to Cart